missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53(phospho S46) & TP53 Protein Phosphorylation Antibody Pair, Abnova™
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the TP53 protein, and the other against the specific S46 phosphorylated site of TP53 for use in in situ Proximity Ligation Assay.
Specifications
Specifications
| Antigen | TP53 |
| Applications | In situ PLA |
| Conjugate | Unconjugated |
| Gene | tumor protein p53 |
| Gene Alias | FLJ92943/LFS1/TRP53/p53 |
| Gene Symbols | TP53 |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 7157 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?