missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TP-040

Product Code. 18785516
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785516 10 mg 10mg
18705536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18785516 Supplier Tocris Bioscience™ Supplier No. 7771/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective O-GlcNAcase (OGA) inhibitor

TP-040 is a potent and selective O-GlcNAcase (OGA) inhibitor (IC50 = 46 nM). It increases the level of O-GlcNAcylated protein in human neuroblastoma SH-SY5Y cells (EC50 = 450 nM). It is suitable for in vivo studies and exhibits brain permeability.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1-Methyl-1 H-imidazol-4-yl)methyl]-4-(4-methyl-1-piperidinyl)-2-pyrimidinamine
CAS 2757254-99-0
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C15H22N6
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.