missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOX3 Monoclonal antibody specifically detects TOX3 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | TOX3 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | TOX3/1124 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CAG trinucleotide repeat-containing gene F9 protein, CAGF9trinucleotide repeat containing 9, TNRC9, TOX high mobility group box family member 3, Trinucleotide repeat-containing gene 9 protein |
| Host Species | Mouse |
| Immunogen | Recombinant fragment of human TOX3 protein (around aa 251-389) (Exact sequence is proprietary) (Uniprot: O15405) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?