missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR/mTOR Antibody (CL8228), Novus Biologicals™
Mouse Monoclonal Antibody
329.00 € - 600.00 €
Specifications
| Antigen | TOR/mTOR |
|---|---|
| Clone | CL8228 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672745
|
Novus Biologicals
NBP3-21165-25ul |
25 μg |
329.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683004
|
Novus Biologicals
NBP3-21165-100ul |
100 μg |
600.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOR/mTOR Monoclonal antibody specifically detects TOR/mTOR in Human samples. It is validated for Western BlotSpecifications
| TOR/mTOR | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| This antibody was developed against a Synthetic Peptide of P42345. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL8228 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, Macroautophagy, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| PBS, pH 7.2, 40% glycerol | |
| 2475 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title