Learn More
TOP2A Monoclonal Antibody (OTI1D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802450S
Description
TOP2A Monoclonal antibody specifically detects TOP2A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TOP2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35968 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3791 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D5 | |
| Unconjugated | |
| KDR | |
| DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase 2-alpha-like protein; DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2; Top-2; Top2a; TOPOII; topoisomerase (DNA) 2 alpha; topoisomerase (DNA) II alpha; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha; topoisomersae II; TP2A | |
| Human recombinant protein fragment corresponding to amino acids 786-1356 of human KDR produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.