Learn More
Bochem™ Tongs for Dishes
Product Code. 10112701
Change view
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10112701 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10112701 Supplier Bochem™ Supplier No. 2280
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Bochem™ Laboratory tongs are scissor-like tools for grasping round bottom dishes.
  • For round bottom dishes approximately 100 to 250 mm diameter
Specifications
Description Tongs for Dishes
For Use With (Application) Grasping Round Bottom Dishes
Length (Metric) 250 mm
Material Stainless Steel (18/10)