missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM40L Mouse anti-Human, Clone: OTI6F3, lyophilized, TrueMAB™
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Potential channel-forming protein implicated in import of protein precursors into mitochondria.
Specifications
Specifications
| Antigen | TOMM40L |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI6F3 |
| Conjugate | Unconjugated |
| Formulation | PBS with 8% trehalose and no preservative; pH 7.3 |
| Gene | TOMM40L |
| Gene Accession No. | Q969M1 |
| Gene Alias | TOMM40B |
| Gene Symbols | TOMM40L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?