missing translation for 'onlineSavingsMsg'
Learn More

Toltrazuril, MedChemExpress

Product Code. p-200101030
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16480798

Brand: Medchem Express HYB0175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Toltrazuril (BAY-i 9142) is an antiprotozoal agent that acts upon Coccidia parasites.

Toltrazuril (BAY-i 9142) is an antiprotozoal agent that acts upon Coccidia parasites.

Specifications

Chemical Name or Material Toltrazuril
Physical Form Solid
CAS 69004-03-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H14F3N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym BAY-i 9142
Solubility Information DMSO : 130 mg/mL (305.61 mM; Need ultrasonic)
SMILES O=C(N(C1=CC=C(OC2=CC=C(SC(F)(F)F)C=C2)C(C)=C1)C(N3)=O)N(C)C3=O
Molecular Weight (g/mol) 425.38
Formula Weight 425.38
Percent Purity 98.65%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.