Learn More
Tofogliflozin (hydrate), MedChemExpress
Product Code. 16485668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485668 10 mg 10mg
16438367 5 mg 5mg
16431837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16485668 Supplier Medchem Express Supplier No. HY13413
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
MedChemExpress Tofogliflozin hydrate (CSG-452 hydrate) is a potent and highly specific sodium/glucose cotransporter 2 (SGLT2) inhibitor with an IC50 of 2.9 nM and Ki values of 2.9 nM, 14.9 nM, and 6.4 nM for human, rat, and mouse SGLT2. Tofogliflozin partially inhibits high glucose-induced reactive oxyen species (ROS) generation in tubular cells.
Tofogliflozin hydrate (CSG-452 hydrate) is a potent and highly specific sodium/glucose cotransporter 2 (SGLT2) inhibitor with an IC50 of 2.9 nM and Ki values of 2.9 nM, 14.9 nM, and 6.4 nM for human, rat, and mouse SGLT2. Tofogliflozin partially inhibits high glucose-induced reactive oxyen species (ROS) generation in tubular cells.
Specifications
Chemical Name or Material Tofogliflozin (hydrate)
CAS 1201913-82-7
Color White
Purity Grade Notes Research
Molecular Formula C22H28O7
Quantity 10 mg
Synonym CSG-452 hydrate
Solubility Information DMSO : ≥ 100 mg/mL (247.25 mM) ∣H2O : 0.33 mg/mL (0.82 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@]21OCC3=C2C=C(CC4=CC=C(CC)C=C4)C=C3.O
Molecular Weight (g/mol) 404.45
Formula Weight 404.45
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism