missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RGDS peptide
GREENER_CHOICE

Product Code. 18024092
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18024092

Brand: Tocris Bioscience™ 3498/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Integrin binding sequence; inhibits integrin receptor function

RGDS peptide is an integrin binding sequence that inhibits integrin receptor function. Decreases systemic inflammation via inhibition of collagen-triggered activation of leukocytes and attenuates expression of inflammatory cytokines, iNOS and MMP-9. Promotes cell attachment and abrogates apoptosis via the mitochondrial pathway in osteoblasts in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 10 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.