Learn More
Tocris Bioscience™ Desmopressin
GREENER_CHOICE
Product Code. 18095962
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18095962
Brand: Tocris Bioscience™ 3396/1
Please to purchase this item. Need a web account? Register with us today!
Synthetic vasopressin analog; V1B and V2 agonist
Desmopressin is a synthetic vasopressin analog that acts as an agonist at V1B and V2 receptors (EC50 values are 11.4 and 23.9 nM and Ki values are 5.84 and 65.9 nM respectively). Prevents polycystic kidney disease formation and exhibits antidiuretic, antiproliferative, hemostatic and hypotensive activity in vivo.
TRUSTED_SUSTAINABILITY
Specifications
Purity 0.95
Quantity 1 mg