missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOCA-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117351
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.
Specifications
| TOCA-1 | |
| Polyclonal | |
| Unconjugated | |
| FNBP1L | |
| 2610318I01Rik; AW548221; C1orf39; Cdc42 effector; FNBP1L; formin binding protein 1 like; formin binding protein 1-like; formin-binding protein 1-like; RGD1305386; TOCA1; Toca-1; transducer of Cdc42-dependent actin assembly 1; transducer of Cdc42-dependent actin assembly protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.8 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q5T0N5 | |
| FNBP1L | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human FNBP1L. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction