Learn More
Hach Lange™ TOC Cuvette Tests
Product Code. 10060274
Click to view available options
No. of Tests:
25
Unit Size:
Each
Product Code. 10060274
Brand: Hach Lange™ LCK381
Please to purchase this item. Need a web account? Register with us today!
Specifications
No. of Tests 25
Range 65 to 735mg/L C
Storage Requirements 15°C to 25°C
Test Method Difference Method (TOC is determined as the difference between the TC and TIC values), Persulphate Digestion
Type TOC Cuvette Test