missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOB1 Polyclonal antibody specifically detects TOB1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TOB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1-5 μg/mL |
| Formulation | Phosphate buffered saline |
| Gene Alias | APRO6, MGC104792, PIG49, protein Tob1, TOBMGC34446, Transducer of erbB-2 1, transducer of ERBB2, 1, TROB, TROB1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the TOB1 |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?