missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNMD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116021
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total tenomodulin.
TNMD is a single-pass type II membrane proteinPotential. It belongs to the chondromodulin-1 family and contains 1 BRICHOS domain. TNMD may be an angiogenesis inhibitor.
Specifications
| TNMD | |
| Polyclonal | |
| Unconjugated | |
| TNMD | |
| 1110017I01Rik; Bricd4; BRICHOS domain containing 4; ChM1L; Chondromodulin-1-like protein; chondromodulin-IB; chondromodulin-I-like protein; hChM1L; hTeM; mChM1L; mTeM; myodulin; rChM1L; rTeM; teM; Tendin; tenomodulin; TNMD; UNQ771/PRO1565; zgc:172161 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64102, 64103, 64104 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9EP64, Q9ESC2, Q9H2S6 | |
| TNMD | |
| A synthesized peptide derived from human TNMD(Accession Q9H2S6), corresponding to amino acid residues R262-R312. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction