missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (6N2E12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27351
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TNF-alpha | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml, Flow Cytometry 0.5-2ug/10^6cells | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 6N2E12 | |
| Unconjugated | |
| P01375 | |
| TNF | |
| Full-length recombinant human TNF alpha was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction