missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM8C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
292.00 € - 559.00 €
Specifications
| Antigen | TMEM8C |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18488451
|
Novus Biologicals
NBP2-34175-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790484
|
Novus Biologicals
NBP2-34175 |
0.1 mL |
559.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM8C Polyclonal specifically detects TMEM8C in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TMEM8C | |
| Polyclonal | |
| Rabbit | |
| Human | |
| A6NI61 | |
| 389827 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KWLQKMKEKKGLYPDKSVYTQQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MYOMAKER, TMEM226, Transmembrane Protein 226, Transmembrane Protein 8C | |
| MYMK | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title