Learn More
Invitrogen™ TMEM59 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140175
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: HLLFPLTLVR SFWSDMMDSA QSFITSSWTF YLQADDGKIV IFQSKPEIQY APHLEQEPTN LRESSLSKMS SDLQMRNSQA HRNFLEDGES DGFLRCLSLN SGW Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 92%.
Processing of the amyloid precursor protein (APP) by two different proteases, called alpha- and beta-secretase, is a central regulatory event in the generation of the amyloid beta peptide (Abeta), which has a key role in Alzheimer disease (AD) pathogenesis. TMEM59, a Golgi-localized protein, modulates the O-glycosylation and complex N-glycosylation steps occurring during the Golgi maturation of several proteins such as APP, BACE1, SEAP or PRNP. It inhibits APP transport and shedding.
Specifications
| TMEM59 | |
| Polyclonal | |
| Unconjugated | |
| TMEM59 | |
| 1110001M20Rik; 3110046P06Rik; AI256529; C1orf8; D4Ertd20e; DCF1; dendritic cell factor 1; HSPC001; Liver membrane-bound protein; MTDCF1; ORF18; PRO195; RGD1310313; RP4-758J24.2; Tdcf1; Thymic dendritic cell-derived factor 1; TMEM59; Transmembrane protein 59; UNQ169; UNQ169/PRO195 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXS4 | |
| TMEM59 | |
| Recombinant protein corresponding to Human TMEM59. Recombinant protein control fragment (Product #RP-109480). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.