missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17095-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in the biosynthesis of the phosphorylated O-mannosyl trisaccharide (N-acetylgalactosamine-beta-3-N-acetylglucosamine-beta-4-(phosphate-6-)mannose), a carbohydrate structure present in alpha-dystroglycan (DAG1), which is required for binding laminin G-like domain-containing extracellular proteins with high affinity.Specifications
| TMEM5 | |
| Polyclonal | |
| Unconjugated | |
| TMEM5 | |
| HP10481, TMEM5, transmembrane protein 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10329, 216395, 299841 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VDX6, Q9Y2B1 | |
| Tmem5 | |
| TMEM5 Fusion Protein Ag10562 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction