Learn More
TLR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NB100-56571SS
This item is not returnable.
View return policy
Description
TLR3 Polyclonal specifically detects TLR3 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Flow (Cell Surface), Flow (Intracellular), Knockdown Validated.
Specifications
| TLR3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Rabbit | |
| 104 kDa | |
| 0.025 mg | |
| Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7098 | |
| Human, Mouse, Pig | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated | |
| O15455 | |
| TLR3 | |
| A mix of synthetic peptides corresponding to amino acids 135-150 (SIHKIKSNPFKNQKNL), 828-844 (CRRFKVHHAVQQAIEQN), and 876-891 (CILNWPVQKERINAFH) of mouse TLR3. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse and Human. Predicted to cross-react with Rat. Porcine data from customer review. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |