missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TLN1 (Human) Recombinant Protein

Human TLN1 partial ORF (AAH42923.1, 1052 a.a. - 1424 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00007094-Q07.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16054895

  • 545.00 € / 25µg

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

  • Sequence: SALSVVQNLEKDLQEVKAAARDGKLKPLPGETMEKCTQDLGNSTKAVSSAIAQLLGEVAQGNENYAGIAARDVAGGLRSLAQAARGVAALTSDPAVQAIVLDTASDVLDKASSLIEEAKKAAGHPGDPESQQRLAQVAKAVTQALNRCVSCLPGQRDVDNALRAVGDASKRLLSDSLPPSTGTFQEAQSRLNEAAAGLNQAATELVQASRGTPQDLARASGRFGQDFSTFLEAGVEMAGQAPSQEDRAQVVSNLKGISMSSSKLLLAAKALSTDPAAPNLKSQLAAAARAVTDSINQLITMCTQQAPGQKECDNALRELETVRELLENPVQPINDMSYFGCLDSVMENSKVLGEAMTGISQNAKNGNLPEFGD
Specifications

Specifications

AAH42923.1
talin 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TLN1
GST
7094
Wheat germ expression system
25 μg
ILWEQ, KIAA1027, TLN
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Promotions

Promotions