missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SDYNLVVDED QPSEPPSPAT TPCGKVPICI PARRDLVDSP ASLASSLGSP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
TLE2 (Transducin Like Enhancer Of Split 2) is a Protein Coding gene. Among its related pathways are Signaling by Wnt and Wnt / Hedgehog / Notch. Gene Ontology (GO) annotations related to this gene include transcription corepressor activity. An important paralog of this gene is TLE1.
Specifications
Specifications
| Antigen | TLE2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TLE2 |
| Gene Accession No. | Q04725 |
| Gene Alias | enhancer of split groucho-like protein 2; ESG; ESG2; ESG-2; GRG2; TLE2; transducin like enhancer of split 2; transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer protein 2 |
| Gene Symbols | TLE2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
