missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TL4-12
GREENER_CHOICE

Product Code. 18784739
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18784739

Brand: Tocris Bioscience™ 5910/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent MAP4K2 (GCK) inhibitor

TL4-12 is a potent MAP4K2 (GCK) inhibitor (IC50 = 37 nM). Exhibits >70-fold selectivity for MAP4K2 over TAK1 (IC50 = 2.7 μM for TAK1). Inhibits IL-1 and TGFβ-induced p38 MAPK phosphorylation, in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methyl-3-[[6-(methylamino)-4-pyrimidinyl]oxy]-N-[3-(4-methyl-1-piperazinyl)-5-(trifluoromethyl)phenyl]benzamide
CAS 1620820-12-3
Quantity 10 mg
Molecular Formula C25H27F3N6O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.