Learn More
Hirschmann™ Titration Unit Suction Check Valve
Description
- For opus™, ceramus™, solarus™ & ceramus™ HF
- With valve ball and seat made from ceramic
- With Pt/Ir valve spring
Specifications
Specifications
| Quantity | 1 Each |
| Type | Suction Valve |
| For Use With (Equipment) | opus™, ceramus™, solarus™, & ceramus™, HF |
| Material | Ceramic Ball and Seat, Pt/Ir Spring |