Learn More
Hirschmann™ Titration Unit Suction Check Valve
Product Code. 10590152
Change view
Click to view available options
:
For Use With (Equipment): opus™, ceramus™, solarus™, & ceramus™, HF
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10590152 For Use With (Equipment): opus™, ceramus™, solarus™, & ceramus™, HF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10590152 Supplier Hirschmann™ Supplier No. 9316528
Only null left
Add to basket
Add to basket
Suction check valve with Pt/Ir spring and ceramic valve ball and seat for ceramus™, solarus™, opus™.
  • For opus™, ceramus™, solarus™ & ceramus™ HF
  • With valve ball and seat made from ceramic
  • With Pt/Ir valve spring
Specifications
Quantity 1 Each
Type Suction Valve
For Use With (Equipment) opus™, ceramus™, solarus™, & ceramus™, HF
Material Ceramic Ball and Seat, Pt/Ir Spring