Learn More
Titanium, Ti 6-2-4-6, R56260, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 6-2-4-6, R56260
Specifications
Specifications
| Chemical Name or Material | Aluminium, Boron, Carbon, Chromium, Cobalt, Copper, Hydrogen, Iron, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Phosphorus, Silicon, Sulfur, Tin, Tungsten, Vanadium, Yttrium, Zirconium |
| Diameter (Metric) Outside | 40 mm |
| Form | Disc |
| Thickness (Metric) | 19 mm |
| Concentration or Composition (by Analyte or Components) | Copper: 29 ppm, Tungsten: 25 ppm, Niobium: 23 ppm, Cobalt: 22 ppm, Sulfur: 21 ppm, Phosphorus: 15 ppm, Boron: 8 ppm, Manganese: 6 ppm, Yttrium: 4 ppm, Nitrogen: 0.0026%, Hydrogen: 0.0036%, Chromium: 0.005%, Vanadium: 0.005%, Carbon: 0.006%, Nickel: 0.007%, Iron: 0.021%, Silicon: 0.052%, Oxygen: 0.114%, Tin: 2%, Zirconium: 4.11%, Aluminium: 5.95%, Molybdenum: 6.04% |