Learn More
Titanium(IV) 2-ethylhexyloxide, 95%, Thermo Scientific™
27.30 € - 256.00 €
Chemical Identifiers
| CAS | 1070-10-6 |
|---|---|
| Molecular Formula | C32H68O4Ti |
| Molecular Weight (g/mol) | 564.76 |
| MDL Number | MFCD00015264,MFCD00015264 |
| InChI Key | KTXWGMUMDPYXNN-UHFFFAOYNA-N |
| Synonym | tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt |
| PubChem CID | 54603245 |
| IUPAC Name | titanium(4+) tetrakis(2-ethylhexan-1-olate) |
| SMILES | [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15492947
|
Thermo Scientific Alfa Aesar
077107.22 |
100g |
27.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15402957
|
Thermo Scientific Alfa Aesar
077107.36 |
500g |
78.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15412957
|
Thermo Scientific Alfa Aesar
077107.A3 |
2kg |
256.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1070-10-6 | |
| 564.76 | |
| KTXWGMUMDPYXNN-UHFFFAOYNA-N | |
| 54603245 | |
| [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-] |
| C32H68O4Ti | |
| MFCD00015264,MFCD00015264 | |
| tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt | |
| titanium(4+) tetrakis(2-ethylhexan-1-olate) |
Specifications
| 1070-10-6 | |
| Ti[OCH2CH(C2H5)(CH2)3CH3]4 | |
| 3692023 | |
| tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt | |
| [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-] | |
| 564.76 | |
| 564.79 | |
| 1.482 | |
| 0.926 g/mL |
| C32H68O4Ti | |
| MFCD00015264,MFCD00015264 | |
| 71°C (160°F) | |
| KTXWGMUMDPYXNN-UHFFFAOYNA-N | |
| titanium(4+) tetrakis(2-ethylhexan-1-olate) | |
| 54603245 | |
| 95% | |
| Moisture sensitive | |
| Titanium (IV) 2-ethylhexyloxide |
Safety and Handling
GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 213-969-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures