Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
Description
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures tissues are safely processed as cassettes will not pop open during processing
Specifications
Specifications
| Pore Style | Mesh Pore Insert |
| Color | Tan |
| Product Type | HistoScreen™ Cassette |