missing translation for 'onlineSavingsMsg'
Learn More

TIP60 Antibody [Janelia Fluor« 525], Novus Biologicals Biologicals™

Product Code. 30498377 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498377 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498377 Supplier Novus Biologicals Supplier No. NBP327886JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIP60 Polyclonal antibody specifically detects TIP60 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP60
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias cPLA2, ESA1EC 2.3.1.48, Histone acetyltransferase HTATIP, histone acetyltransferase KAT5, HIV-1 Tat interactive protein, HIV-1 Tat interactive protein, 60kDa, HTATIP1, HTATIPcPLA2 interacting protein, K(lysine) acetyltransferase 5,60 kDa Tat-interactive protein, K-acetyltransferase 5, Lysine acetyltransferase 5, PLIPTIP, Tat interacting protein, 60kDa, Tip60, TIP60cPLA(2)-interacting protein
Host Species Rabbit
Immunogen TIP60 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human TIP60. The immunogen is located within the last 50 amino acids of TIP60.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 10524
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.