Learn More
Tioclomarol, TRC
Specifications
Specifications
| Chemical Name or Material | Tioclomarol |
| CAS | 22619-35-8 |
| Molecular Formula | C22 H16 Cl2 O4 S |
| Quantity | 10 mg |
| Synonym | 2H-1-Benzopyran-2-one','3-[3-(4-chlorophenyl)-1-(5-chloro-2-thienyl)-3-hydroxypropyl]-4-hydroxy-, Coumarin','3-[5-chloro-α-(p-chloro-β-hydroxyphenethyl)-2-thenyl]-4-hydroxy- (8CI), 3-[3-(4-Chlorophenyl)-1-(5-chloro-2-thienyl)-3-hydroxypropyl]-4-hydroxy-2H-1-benzopyran-2-one, Apegmone, Tioclomarol |
| InChI Formula | InChI=1S/C22H16Cl2O4S/c23-13-7-5-12(6-8-13)16(25)11-15(18-9-10-19(24)29-18)20-21(26)14-3-1-2-4-17(14)28-22(20)27/h1-10,15-16,25-26H,11H2 |
| SMILES | OC(CC(C1=C(O)c2ccccc2OC1=O)c3ccc(Cl)s3)c4ccc(Cl)cc4 |
| IUPAC Name | 3-[3-(4-chlorophenyl)-1-(5-chlorothiophen-2-yl)-3-hydroxypropyl]-4-hydroxychromen-2-one |
| Molecular Weight (g/mol) | 447.33 |
| Formula Weight | 446.0146 |