Learn More
Lovibond™ Tintometer™ Phenol Red Solution, Refil Bottle, 100mL
Product Code. 15342015
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15342015 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15342015 Supplier Lovibond™ Supplier No. 471041
Please to purchase this item. Need a web account? Register with us today!
For use with Lovibond test kits, colorimeters and spectrophotometers
  • Phenol Red solution
  • pH indicator
  • Refil bottle
Specifications
For Use With (Application) pH Test
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 mL
Type Phenol Red Solution Refil Bottle
compliance-icons
Product Identifier
  • PHENOLRED SOLUTION - REFILL BOTTLE .
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P233-Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: butan-2-ol