missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ DPD 3 Reagent Solution, 15mL
Description
For use with Lovibond test kits, colorimeters and spectrophotometers
- DPD 3 Solution
- Black Bottle
- Red Cap
- 15mL
- Pack of 6 bottles
Specifications
Specifications
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 6 x 15 mL Bottles |
| Type | DPD 3 Reagent Solution |
Safety and Handling
Product Identifier
- 15ML DPD 3 SOLUTION, PACK OF 6 .
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contain: potassium iodide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?