missing translation for 'onlineSavingsMsg'
Learn More

Tinidazole, MedChemExpress

Product Code. 16400808
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
This item is not returnable. View return policy

Product Code. 16400808

Brand: Medchem Express HYB0177

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa.

Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa.

Specifications

Chemical Name or Material Tinidazole
Physical Form Solid
CAS 19387-91-8
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H13N3O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (202.21 mM) ∣H2O : 3.33 mg/mL (13.47 mM; Need ultrasonic)
SMILES CC1=NC=C([N+]([O-])=O)N1CCS(=O)(CC)=O
Molecular Weight (g/mol) 247.27
Formula Weight 247.27
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.