missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-1 Antibody (63515) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB970H
This item is not returnable.
View return policy
Description
TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| TIMP-1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 63515 | |
| Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-1, Cys24-Ala207, Accession # P01033 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix | |
| 7076 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction