Learn More
Timosaponin BII, MedChemExpress
Product Code. 16451518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451518 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16451518 Supplier Medchem Express Supplier No. HYN0812
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
MedChemExpress Timosaponin BII (Prototimosaponin A III) is a steroid saponin found in the rhizomes of Anemarrhena asphodeloides. Timosaponin BII has neuronal protective, anti-inflammatory and antioxidant activities.
Timosaponin BII (Prototimosaponin A III) is a steroid saponin found in the rhizomes of Anemarrhena asphodeloides. Timosaponin BII has neuronal protective, anti-inflammatory and antioxidant activities.
Specifications
Chemical Name or Material Timosaponin BII
CAS 136656-07-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C45H76O19
Quantity 5 mg
Synonym Prototimosaponin A III
Solubility Information DMSO : 100 mg/mL (108.57 mM; Need ultrasonic) ∣H2O : 100 mg/mL (108.57 mM; Need ultrasonic)
SMILES C[C@@]1([C@]([C@@H]2C)([H])[C@](O[C@]2(O)CC[C@H](C)CO[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO)([H])C4)[C@]4([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC1
Molecular Weight (g/mol) 921.07
Formula Weight 921.07
Percent Purity 98.52%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration