Learn More
Tilorone Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Tilorone Dihydrochloride |
| CAS | 27591-69-1 |
| Molecular Formula | C25 H34 N2 O3 . 2 Cl H |
| Quantity | 50 mg |
| Synonym | 9H-Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','hydrochloride (1:2), 9H-Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','dihydrochloride (9CI), Fluoren-9-one','2,7-bis[2-(diethylamino)ethoxy]-','dihydrochloride (8CI), Amixinum, Amyxin, DEAE-F, NSC 143969, RMI 10008DA, RMI 10028DA, Tilorone dihydrochloride, Tilorone hydrochloride |
| InChI Formula | InChI=1S/C25H34N2O3.2ClH/c1-5-26(6-2)13-15-29-19-9-11-21-22-12-10-20(30-16-14-27(7-3)8-4)18-24(22)25(28)23(21)17-19;;/h9-12,17-18H,5-8,13-16H2,1-4H3;2*1H |
| SMILES | Cl.Cl.CCN(CC)CCOc1ccc2c(c1)C(=O)c3cc(OCCN(CC)CC)ccc23 |
| IUPAC Name | 2,7-bis[2-(diethylamino)ethoxy]fluoren-9-one;dihydrochloride |
| Molecular Weight (g/mol) | 483.47 |
| Formula Weight | 482.2103 |