Learn More
Tocris Bioscience™ Tilfrinib
Description
Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.
Specifications
Specifications
| Chemical Name or Material | 3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol |
| CAS | 1600515-49-8 |
| Quantity | 10 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C17H13N3O |
| Formula Weight | 275.3 |
| Purity | >98% |