Learn More
Tigecycline, MedChemExpress
Product Code. 16498106
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Product Code. 16498106
Brand: Medchem Express HYB0117
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tigecycline (GAR-936) is a broad-spectrum glycylcycline antibiotic. The mean inhibitory concentration (MIC) of Tigecycline for E. coli (MG1655 strain) is approximately 125 ng/mL. MIC50 and MIC90 are 1 and 2 mg/L for Acinetobacter baumannii (A. baumannii), respectively.
Tigecycline (GAR-936) is a broad-spectrum glycylcycline antibiotic. The mean inhibitory concentration (MIC) of Tigecycline for E. coli (MG1655 strain) is approximately 125 ng/mL. MIC50 and MIC90 are 1 and 2 mg/L for Acinetobacter baumannii (A. baumannii), respectively.
Specifications
Chemical Name or Material Tigecycline
Physical Form Solid
CAS 220620-09-7
Color Brownish Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H39N5O8
For Use With (Application) COVID-19-immunoregulation
Synonym GAR-936
Solubility Information DMSO : 150 mg/mL (256.13 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (14.22 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])CC4=C(C(C3=C(O)[C@@]21O)=O)C(O)=C(NC(CNC(C)(C)C)=O)C=C4N(C)C)N
Molecular Weight (g/mol) 585.65
Formula Weight 585.65
Percent Purity 98.03%
Grade Research