missing translation for 'onlineSavingsMsg'
Learn More

Bandelin Electronic™ Tickopur RW 77 Special Cleaners

Product Code. 10485421
Click to view available options
Volume (Metric):
1 L
2 L
5 L
Unit Size:
1 L
2 L
5 L
This item is not returnable. View return policy

Product Code. 10485421

Brand: Bandelin Electronic™ 898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Special Alkaline Cleaner
Form Liquid
pH 9.9
Product Line Tickopur
Appearance Clear Pale Yellow
Container Type Polyethylene Henkel bottle
For Use With (Application) For Cleaning of disassembled and undivided Machinery and tool parts, printed circuit boards in the service area, clocks, test sieves, coins
Quantity 1 Ea.
Volume (Metric) 2 L
compliance-icons
Product Identifier
  • Tickopur RW77
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : C16-C18 Fatty acid TEA, 2-propanol, C13-C17 alkansulfonate,ammoniac
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.