missing translation for 'onlineSavingsMsg'
Learn More

Bandelin Electronic™ Tickopur R 32 Special Cleaner

Product Code. 10780693
Change view
Click to view available options
Volume (Metric):
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
10780693 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10780693 Supplier Bandelin Electronic™ Supplier No. 882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Special Cleaner
Form Liquid
pH 9.9
Product Line Tickopur
Appearance Colorless
Container Type Polyethylene Henkel bottle
For Use With (Application) Cleaning of disassembled and undivided Machinery, tools, equipment and parts specially in electroplating, analytics and Laser Technology
Quantity 1 Ea.
Volume (Metric) 2 L
compliance-icons
Product Identifier
  • Tickopur R32
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: phosphoric acid partial ester sodium salt, Dinatriummetasilikat-pentahydrat, C8-C18 Aminoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.