Learn More
Ticagrelor Sulfone, TRC
Specifications
Specifications
| Chemical Name or Material | Ticagrelor Sulfone |
| CAS | 274693-39-9 |
| Molecular Formula | C23H28F2N6O6S |
| Quantity | 10 mg |
| Synonym | Ticagrelor Impurity M, (1S,2S,3R,5S)-3-(7-(((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-(propylsulfonyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-5-(2-hydroxyethoxy)cyclopentane-1,2-diol |
| InChI Formula | InChI=1S/C23H28F2N6O6S/c1-2-7-38(35,36)23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(37-6-5-32)20(34)19(16)33/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16+,17-,19-,20+/m0/s1 |
| SMILES | CCCS(=O)(=O)c1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCO)[C@@H](O)[C@H]5O)c4n1 |
| IUPAC Name | (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfonyltriazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentane-1,2-diol |
| Molecular Weight (g/mol) | 554.57 |
| Formula Weight | 554.1759 |