missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticagrelor, MedChemExpress
Description
Ticagrelor (AZD6140) is a reversible oral P2Y12 receptor antagonist for the treatment of platelet aggregation.
Specifications
Specifications
| Chemical Name or Material | Ticagrelor |
| CAS | 274693-27-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H28F2N6O4S |
| Quantity | 10 mg |
| Synonym | AZD6140 AR-C 126532XX |
| Solubility Information | DMSO : ≥ 50 mg/mL (95.68 mM) |
| SMILES | O[C@H]1[C@@H](O)[C@H](N2N=NC3=C(N[C@H]4[C@H](C5=CC=C(F)C(F)=C5)C4)N=C(SCCC)N=C32)C[C@@H]1OCCO |
| Molecular Weight (g/mol) | 522.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?