Learn More
Ticagrelor, MedChemExpress
Product Code. 16493108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493108 10 mg 10mg
16459666 50 mg 50mg
16444406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16493108 Supplier Medchem Express Supplier No. HY10064
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ticagrelor (AZD6140) is a reversible oral P2Y12 receptor antagonist for the treatment of platelet aggregation.
Ticagrelor (AZD6140) is a reversible oral P2Y12 receptor antagonist for the treatment of platelet aggregation.
Specifications
Chemical Name or Material Ticagrelor
CAS 274693-27-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H28F2N6O4S
Quantity 10 mg
Synonym AZD6140 AR-C 126532XX
Solubility Information DMSO : ≥ 50 mg/mL (95.68 mM)
SMILES O[C@H]1[C@@H](O)[C@H](N2N=NC3=C(N[C@H]4[C@H](C5=CC=C(F)C(F)=C5)C4)N=C(SCCC)N=C32)C[C@@H]1OCCO
Molecular Weight (g/mol) 522.57
Formula Weight 522.57
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation