Learn More
Ticagrelor Acetonide, TRC
Product Code. 30379950
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379950 1 g
30372381 100 mg
30372167 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30379950 Supplier Toronto Research Chemicals Supplier No. TRCD4453051G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 274693-26-4
Molecular Formula C26 H32 F2 N6 O4 S
Molecular Weight (g/mol) 562.63
Synonym 2-[[(3aR,4S,6R,6aS)-6-[7-[[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-3-yl]tetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-yl]oxy]ethanol, 2-[[(3aS,4R,6S,6aa)-4-[7-[[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl]-2,2-dimethyl-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]oxy]ethanol
IUPAC Name 2-[[(3aR,4S,6R,6aS)-6-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-3-yl]-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl]oxy]ethanol
SMILES CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCO)[C@H]6OC(C)(C)O[C@@H]56)c4n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C26H32F2N6O4S/c1-4-9-39-25-30-23(29-17-11-14(17)13-5-6-15(27)16(28)10-13)20-24(31-25)34(33-32-20)18-12-19(36-8-7-35)22-21(18)37-26(2,3)38-22/h5-6,10,14,17-19,21-22,35H,4,7-9,11-12H2,1-3H3,(H,29,30,31)/t14-,17+,18+,19-,21-,22+/m0/s1
Formula Weight 562.2174
Chemical Name or Material 2-[{(3aS,4R,6S,6aa)-4-[7-[{(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl}amino]-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl]-2,2-dimethyl-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl}oxy]ethanol