Learn More
Tocris Bioscience™ Tiagabine hydrochloride
GREENER_CHOICE
Product Code. 18054092
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18054092
Brand: Tocris Bioscience™ 4256/10
Please to purchase this item. Need a web account? Register with us today!
Selective GAT-1 inhibitor; anticonvulsant
GABA uptake inhibitor (IC50 = 67 nM in vivo). Exhibits high affinity and selectivity for the GAT-1 GABA transporter. Anticonvulsant; also attenuates established dynorphin-induced allodynia in a mouse model after systemic administration.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145821-59-6
Molecular Formula C20H26ClNO2S2
Specifications
Quantity 10 mg
Formula Weight 412.01
Purity >99%
Molecular Weight (g/mol) 412.00
Chemical Name or Material Tiagabine hydrochloride