missing translation for 'onlineSavingsMsg'
Learn More

Thymidine 5’-Diphosphate Sodium Salt (~90%), Thymidine 5’-Diphosphate Sodium Salt (~90%), TRC

Product Code. 30010463
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010463 10 mg 10mg
30005885 1 mg 1mg
30004314 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010463 Supplier Toronto Research Chemicals Supplier No. TRCT41201010MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 H15 N2 O11 P2 . Na
Molecular Weight (g/mol) 424.17
Synonym Thymidine 5'-Diphosphate Sodium Salt
IUPAC Name sodium;[hydroxy-[[(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy]phosphoryl] hydrogen phosphate
SMILES [Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP(=O)(O)OP(=O)(O)[O-])O2)C(=O)NC1=O

Specifications

CAS 108322-12-9
Quantity 10 mg
InChI Formula InChI=1S/C10H16N2O11P2.Na/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(22-8)4-21-25(19,20)23-24(16,17)18;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,11,14,15)(H2,16,17,18);/q;+1/p-1/t6-,7+,8+;/m0./s1
Formula Weight 424.0049
Chemical Name or Material Thymidine 5'-diphosphate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.