Learn More
Honeywell™ Thunder T1Fs Folding Earmuff
Description
- Optimal attenuation across all frequencies, without increasing earcup size or weight
- Patented baseplate chamber and high-tech non-woven layer manage the flow of air inside the earmuff to control how sound reaches the ear
- Dialectric/plastic construction for durability and protection in electrical environments
- Folds down for compact, convenient storage
- Non-deforming out headband
- Quick click height adjustment
- Belt case available separately
Specifications
Specifications
| Color | Black, Green, Gray |
| Certifications/Compliance | EC Category PPE: 2, QA: ISO 900//2000, EC Certificate: 0401097, EC Attestation: 200422139 |
| Noise Reduction Rating (NRR) | SNR: 31 db |
| Dielectric | Yes |
| Material | Plastic (or dielectric), POM, PC/PBT, PP, PUR-E, PVC |
| Style | Folding |
| Foldable | Yes |