missing translation for 'onlineSavingsMsg'
Learn More

THS-044, MedChemExpress

Product Code. p-200101732
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453777

Brand: Medchem Express HY19621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress THS-044 binding stabilizes the HIF2(alpha) PAS-B folded state, for regulating HIF2 activity in endogenous and clinical settings.

THS-044 binding stabilizes the HIF2(alpha) PAS-B folded state, for regulating HIF2 activity in endogenous and clinical settings.

Specifications

Chemical Name or Material THS-044
Physical Form Powder
CAS 62054-67-5
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H12F3N3O3
For Use With (Application) Cancer-programmed cell death
SMILES FC(C1=CC=C(NN2CCOCC2)C([N+]([O-])=O)=C1)(F)F
Molecular Weight (g/mol) 291.23
Formula Weight 291.23
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.