missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRSP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13054-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody works well in IHC, WB and IF.
The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism.Specifications
| THRSP | |
| Polyclonal | |
| Unconjugated | |
| THRSP | |
| S14, Spot 14 protein, SPOT14, THRSP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21835, 25357, 7069 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04143, Q62264, Q92748 | |
| THRSP | |
| THRSP Fusion Protein Ag3721 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction