missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42213PCP
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Mouse | |
| 60.3 kDa | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4C in the dark. | |
| IgG1 λ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6C8.2F9 | |
| Flow Cytometry | |
| P07204 | |
| THBD | |
| Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction