Learn More
Gerresheimer™ Threaded Narrow Mouth Glass Bottles without Closure
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Packungsgröße:
Each
Pack of 120
Pack of 35
Pack of 42
Beschreibung
- Brown or clear glass
- Narrow mouth
- Closures sold separately
Spezifikation
Spezifikation
| Capacity (Metric) | 100 mL |
| Color | Clear |
| Material | Borosilicate Glass |
| Product Line | Bottles |
| Closure Included | No |
| Mouth | Narrow |
| Closure Type | Not Included |